Isocianato de 4-fluorobencilo, 98 %, Thermo Scientific Chemicals
Isocianato de 4-fluorobencilo, 98 %, Thermo Scientific Chemicals
Isocianato de 4-fluorobencilo, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isocianato de 4-fluorobencilo, 98 %, Thermo Scientific Chemicals

CAS: 132740-43-3 | C8H6FNO | 151.14 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Envase:
Frasco de vidrio
Número de catálogo 319300010
Precio (USD)
-
Cantidad:
1 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS132740-43-3
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5040 to 1.5060 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only