Nalgene Torque Fitting Tool for Nalgene Bottle and Closure Combinations
Nalgene Torque Fitting Tool for Nalgene Bottle and Closure Combinations
Nalgene Torque Fitting Tool for Nalgene Bottle and Closure Combinations
Thermo Scientific™
Nalgene Torque Fitting Tool for Nalgene Bottle and Closure Combinations
Ensure a guaranteed leak-proof seal with proper torque settings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
3195-043838-430 Closures
3195-001313 mm Closures
3195-002020 mm Closures
3195-002424 mm Closures
3195-002828 mm Closures
3195-003838-415 Closures
3195-004343 mm Closures
3195-004848 mm Closures
3195-005353 mm Closures
3195-006363 mm Closures
3195-015353B Closures
3195-018383B Closures
3195-102120 mm Serum Vial Closures, HDPE
3195-303838 mm Biotainer Closures
3195-904848 mm Biotainer Closures
Catalog number 3195-0438
Price (MXN)
-
For Use With (Equipment):
38-430 Closures
The Nalgene Bottle Torque Fittings represent a specialized closure solution engineered to deliver precise torque application and guaranteed leakproof performance across Nalgene's laboratory vessel portfolio. These fittings are uniquely designed for compatibility with various Nalgene bottle and closure configurations, including lab bottles, packaging bottles, media bottles, biotainers, and PETG serum vials.
  • Comprehensive Size Range: Available in multiple sizes engineered to accommodate the majority of standard laboratory bottles and closure configurations
  • Multi-Material Closure Compatibility: Designed for optimal performance with both high-density polyethylene (HDPE) and polypropylene (PP) closure materials
  • Nalgene-Specific Design: Custom-engineered geometry specifically optimized for Nalgene laboratory closure systems
  • Universal Torque Tool Integration: Compatible with industry-standard 1/4 in. and 3/8 in. square drive torque tools for precise torque application
  • Manual and Automated Operation: Supports both manual torque application and automated torque systems for laboratory workflow integration
  • Product Specifications and Operating Parameters - Detailed torque settings are available in the documents section of this page.
  • Comprehensive Documentation Provides Essential Operating Procedures - Detailed operational instructions are provided in the accompanying documentation section to ensure proper implementation and use of the bottle and closure system.
Specifications
For Use With (Application)Proper torque of closures
For Use With (Equipment)38-430 Closures
Product TypeTorque Fitting
Unit SizeCase of 1