Search
View additional product information for Goat anti-Mouse IgG2a Fc gamma Cross Adsorbed Secondary Antibody, APC - FAQs (31983)
1 product FAQs found
If the two primary antibodies are from different isotypes (such as mouse IgG1 and IgG2a), there are isotype-specific secondary antibodies to choose from. The other option is to make direct conjugates of the primary antibodies through the use of antibody label kits, reactive dyes, or custom conjugations.