Miser™ Antibody Extender Solution NC

Catalog number: 32110B

Thermo Scientific™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Antibody Extender Solution NC is a post-transfer nitrocellulose membrane-treatment reagent that enables Western blot detection with much less primary antibody.

Features of Antibody Extender Solution NC:

Achieve equivalent signal—use at least three times less primary antibody than normal and retain comparable or better signal detectability
Any detection mode or enzyme—the method works with both colorimetric and chemiluminescent detection modes and with horseradish peroxidase (HRP) and alkaline phosphatase (AP) systems
Easy-to-perform, 10-minute protocol—invest a few minutes and save money on your primary antibody; our scientists have reduced their use of expensive primary antibodies by up to 100-fold with this product

By treating nitrocellulose membranes for a few minutes in this reagent immediately after transfer and before probing, you can use much less (more dilute) primary antibody solutions while obtaining the same level of detection. This conserves use of expensive and rare antibodies, enabling more experiments to be performed with the same amount of antibody. The reagent is so effective that our scientists have observed antibody cost reductions of 10-, 25- and even 100-times (depending on the antigen and primary antibody directed against that antigen).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Line: Miser™
Product Size: Custom
Reagent Type: Signal Enhancers, Stabilizing Reagent
Validated Application: ELISA, Western Blot

Contents & storage

Store at room temperature.

Documents

Manuals & protocols