Metanol d4, para RMN, 99,8% de átomo D, Thermo Scientific Chemicals
Metanol d4, para RMN, 99,8% de átomo D, Thermo Scientific Chemicals
Metanol d4, para RMN, 99,8% de átomo D, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Metanol d4, para RMN, 99,8% de átomo D, Thermo Scientific Chemicals

CAS: 811-98-3 | CH4O | 36.066 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
10 ml
25mL.
100 g
Envase:
Frasco de vidrio
Número de catálogo 321280050
Precio (EUR)
-
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS811-98-3
IUPAC Name(²H₃)methan(²H)ol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-MZCSYVLQSA-N
SMILES[2H]OC([2H])([2H])[2H]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Water=<0.05 % (Coulometric)
D-Enrichment>=99.75 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Para RMN

RUO – Research Use Only