Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Tray Pack
Thermo Scientific™

Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Tray Pack

Safely store and ship small-volume samples and valuable reagents with these PPCO diagnostic bottles, for use in test kits and many small-volume applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Neck Finish
322006-905015 mL20-415 mm
322006-91254 mL13-415 mm
322006-90258 mL20-415 mm
Catalog number 322006-9050
Price (EUR)
1.589,00
Case of 1500
Capacity (Metric):
15 mL
Neck Finish:
20-415 mm
Price (EUR)
1.589,00
Case of 1500

Safely store and ship small-volume samples and valuable reagents. Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closures are used in test kits and many small-volume applications. These autoclavable and durable polypropylene copolymer bottles are tray packed with closure for easy handling and feature uniform walls resistant to splitting or puncturing.

Features:
  • Nonsterile
  • Natural translucent PPCO with Natural PP Closure
  • Leakproof† when used with Nalgene closures

Ordering Information:

Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
AutoclavableAutoclavable
Capacity0.5 oz., 15 mL
Capacity (English)0.5 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure MaterialPolypropylene
Closure Size20-415 mm
Closure TypeScrew Cap
DescriptionNatural polypropylene w/PP Closures; Non-sterile; 15mL
Diameter (Metric)25 mm
Diameter (Metric) Inner Neck13 mm
Height (English)2.28 in.
Height (Metric)58 mm
Neck Finish20-415 mm
No. per Case1500
No. per Pack150
Product TypeSample Container
ShapeRound
Width (English)0.98 in.
Capacity (Metric)15 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialPPCO
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 1500

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.