Nalgene™ Square Amber PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
Nalgene™ Square Amber PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
Nalgene™ Square Amber PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
Nalgene™ Square Amber PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
Thermo Scientific™

Nalgene™ Square Amber PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays

Safely store light-sensitive medias, buffers and sera in these square, amber PETG bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
322021-003030 mL20 mm
322021-006060 mL24 mm
322021-0125125 mL38-430 mm
Catalog number 322021-0030
Price (CLP)
-
Capacity (Metric):
30 mL
Closure Size:
20 mm

Thermo Scientific™ Nalgene Square Amber PETG Media Bottles include HDPE closures. Translucent amber bottles meet USP light-resistant requirements and are ideal for medias, buffers and sera.

  • Heavy-walled, shatter resistant and durable
  • Space-saving square design
  • Molded in graduations
  • Nonsterile, non-pyrogenic, non-cytotoxic
  • Leakproof†, with drip-proof neck finish and molded-in neck ring
  • PETG provides excellent gas barrier, with transparency and reduced permeability to CO2 and O2
  • Amber bottle and closure provide protection for light sensitive reagents

Ordering Information:

Products are shrink-wrapped into trays, then packed in double PE lined cartons for cleanroom applications.

Recommended for:

For storage and shipping of light sensitive liquid media, buffers, and sera

Note:

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity (English)1 oz.
Certifications/CompliancePETG Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI.
Closure MaterialHDPE
Closure Size20 mm
Diameter (English)1.5 in.
Diameter (English) Inner Neck0.55 in.
Diameter (Metric)38 mm
Diameter (Metric) Inner Neck14 mm
Height (English)2.5 in.
Height (Metric)64 mm
No. per Case280
No. per Pack40
Product TypeMedia Bottle
ShapeSquare
Capacity (Metric)30 mL
Closure IncludedYes
ColorAmber
GraduatedYes
MaterialPETG
MouthWide
SterilityNon-sterile
Unit SizeCase of 280