Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
Actual product may vary
Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules

Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules

Protect against leakage, breakage and contamination by using PETG nonsterile serum vials with continuous thread closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
322032-00055 mL
322032-001010 mL
322032-002020 mL
Catalog number 322032-0005
HSN 39233090
Price (INR)
108,287.00
Case
Volume (Metric):
5 mL
Price (INR)
108,287.00
Case

Protect against leakage, breakage, and contamination using Thermo Scientific™ Nalgene Nonsterile Serum Vials. Lighter than glass. Use with lined continuous thread closures (available separately).

  • PETG
  • Non-cytotoxic
  • Non-pyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Nonsterile
  • Leakproof† with Nalgene lined continuous thread closures (available separately)

Ordering Information:

Serum Vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 312158).

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
DescriptionSerum Vials
Diameter (Metric) Inner11 mm
ColorClear
Diameter (Metric) Outer22 mm
MaterialPETG
SterilityNon-sterile
Volume (Metric)5 mL
No. per Case1656
No. per Pack276
Volume (English)1/6 oz.
Product LineNalgene
Volume (Metric) Total5 mL
Closure TypeScrew Cap (Not Included)
Quantity276 Pk. , 1656 Cases
Closure Size20-415
For Use With (Equipment)Lined Continuous Thread Closure
Certifications/ComplianceISO 13485:2003
Height (Metric)44 mm
Product TypeSerum Vial
Unit SizeCase