Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
Thermo Scientific™

Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules

Protect against leakage, breakage and contamination by using PETG nonsterile serum vials with continuous thread closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
322032-002020 mL
322032-00055 mL
322032-001010 mL
Catalog number 322032-0020
Price (USD)
244.71
Online Exclusive
387.09
Save 142.38 (37%)
Case of 612
Volume (Metric):
20 mL
Price (USD)
244.71
Online Exclusive
387.09
Save 142.38 (37%)
Case of 612

Protect against leakage, breakage, and contamination using Thermo Scientific™ Nalgene Nonsterile Serum Vials. Lighter than glass. Use with lined continuous thread closures (available separately).

  • PETG
  • Non-cytotoxic
  • Non-pyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Nonsterile
  • Leakproof† with Nalgene lined continuous thread closures (available separately)

Ordering Information:

Serum Vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 312158).

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Certifications/ComplianceISO 13485:2003
Closure Size20-415
DescriptionSerum Vials
Diameter (Metric) Inner11 mm
For Use With (Equipment)Lined Continuous Thread Closure
Height (Metric)64 mm
No. per Case612
No. per Pack153
Product TypeSerum Vial
Volume (English)2/3 oz.
Volume (Metric) Total20 mL
Closure TypeContinuous-thread Closure (Sold Separately)
ColorClear
Diameter (Metric) Outer30 mm
MaterialPETG
Product LineNalgene
Quantity153 Pk., 612 Cases
SterilityNon-sterile
Volume (Metric)20 mL
Unit SizeCase of 612