Potassium dihydrogen phosphate, 98.0-100.5% (dried basis), crystals, NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Potassium dihydrogen phosphate, 98.0-100.5% (dried basis), crystals, NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets NF, BP and EP specifications.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 g
500 mL
2.5 kg
2.5 L
7 lb.
12 kg
385 lb.
Verpackung:
Alle anzeigen
Amber glass bottle, wide mouth, tamper-evident cap
Katalognummer 3248-01
Preis (EUR)
-
Menge:
500 g
Verpackung:
Amber glass bottle, wide mouth, tamper-evident cap
This product is GMP Manufactured and meets NF, BP and EP specifications.
This product is GMP Manufactured and meets NF, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
FarbeWeiß
Schmelzpunkt253°C
VerpackungAmber glass bottle, wide mouth, tamper-evident cap
Siedepunkt∼100°C (1013 hPa)
GüteMulti-Compendial, NF
Molekulargewicht (g/mol)136.09
Reinheit (%)98 to 100.5%
Physikalische FormSolid
Menge500 g
pH4.2 to 4.5
Unit SizeEach