Potassium dihydrogen phosphate, 98.0-100.5% (dried basis), crystals, NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Potassium dihydrogen phosphate, 98.0-100.5% (dried basis), crystals, NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets NF, BP and EP specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
500 mL
2.5 kg
2.5 L
7 lb.
12 kg
385 lb.
Envase:
Ver todo
Polyethylene Pail with Screw Top
Número de catálogo 3248-07
Precio (MXN)
-
Cantidad:
12 kg
Envase:
Polyethylene Pail with Screw Top
This product is GMP Manufactured and meets NF, BP and EP specifications.
This product is GMP Manufactured and meets NF, BP and EP specifications.
Especificaciones
ColorBlanco
Punto de fusión253°C
EnvasePolyethylene Pail with Screw Top
Punto de ebullición∼100°C (1013 hPa)
GradoMulti-Compendial, NF
Peso molecular (g/mol)136.09
Porcentaje de pureza98 to 100.5%
Forma físicaSolid
Cantidad12 kg
pH4.2 to 4.5
Unit SizeEach