Potassium dihydrogen phosphate, 98.0-100.5% (dried basis), crystals, NF, Multi-Compendial, GMP, J.T.Baker™
Potassium dihydrogen phosphate, 98.0-100.5% (dried basis), crystals, NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Potassium dihydrogen phosphate, 98.0-100.5% (dried basis), crystals, NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets NF, BP and EP specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
500 mL
2.5 kg
2.5 L
7 lb.
12 kg
385 lb.
Envase:
Ver todo
Poly bottle, tamper-evident cap
Amber glass bottle, wide mouth
Número de catálogo 3248-15
Precio (USD)
-
Solicite un presupuesto
Cantidad:
2.5 kg
Envase:
Poly bottle, tamper-evident cap
This product is GMP Manufactured and meets NF, BP and EP specifications.
This product is GMP Manufactured and meets NF, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
ColorBlanco
Punto de fusión253°C
EnvasePoly bottle, tamper-evident cap
Punto de ebullición∼100°C (1013 hPa)
GradoMulti-Compendial, NF
Peso molecular (g/mol)136.09
Porcentaje de pureza98 to 100.5%
Forma físicaSolid
Cantidad2.5 kg
pH4.2 to 4.5
Unit SizeEach