Dipotassium hydrogen phosphate, 98.0-100.5% (dried basis),USP, Multi-Compendial, Endotoxin-free, GMP, J.T.Baker™
Avantor™

Dipotassium hydrogen phosphate, 98.0-100.5% (dried basis),USP, Multi-Compendial, Endotoxin-free, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
5 kg
12 kg
Catalog number 3250-01
Price (MXN)
-
Quantity:
500 g
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorWhite
Melting Point340°C
PackagingPoly bottle, tamper-evident cap
GradeMulti-Compendial/USP
Molecular Weight (g/mol)174.18
Percent Purity98 to 100.5%
Physical FormSolid
Quantity500 g
pH8.5 to 9.6
Unit SizeEach