William's E Medium, GlutaMAX™ Supplement
Gibco™

William's E Medium, GlutaMAX™ Supplement

GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
32551020500 mL
32551087
32551-087으로도 사용됨
10 x 500 mL
카탈로그 번호 32551020
제품 가격(KRW)
124,000
온라인 행사
Ends: 31-Dec-2026
137,000
할인액 13,000
Each
수량:
500 mL
Customize this product
제품 가격(KRW)
124,000
온라인 행사
Ends: 31-Dec-2026
137,000
할인액 13,000
Each
William's E Medium was originally developed by Williams and Gunn as reduced serum-supplemented medium for long-term cell cultures of adult rat liver epithelial cells. Gibco™ William's E Medium can also be used for growing hepatocyte cells of different species, such as human derived HepaRG™ cells.

This William's E Medium is modified as follows:

  • With: Phenol Red, GlutaMAX™
  • Without: HEPES

The complete formulation is available.

Gibco™ William's E Medium is a modification of earlier media that has been enriched in amino acids and double the glucose. William's E Medium contains unique ingredients, including zinc, iron, manganese, non-essential amino acids, the reducing agent glutathione and the lipid methyl linoleate. Gibco™ William's E Medium with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

cGMP Manufacturing

Gibco™ William's E Medium is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

William's E Medium contains no proteins or growth factors. Therefore, William's E Medium requires supplementation, commonly with 5-10% Fetal Bovine Serum (FBS). William's E Medium uses a sodium bicarbonate buffer system (2.2 g / L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

사양
세포 유형Mammalian Cells
제품라인GlutaMAX
제품 유형William's E Medium
수량500 mL
유통 기한12 Months From Date of Manufacture
배송 조건Room Temperature
분류Animal Origin-free, Serum-free, Protein-free
Culture TypeMammalian Cell Culture
형태Liquid
Serum LevelSerum-free
멸균Sterile-filtered
Sterilization MethodSterile-filtered
첨가제 포함GlutaMAX, Phenol Red
첨가제 없음No HEPES
Unit SizeEach
구성 및 보관
Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

자주 묻는 질문(FAQ)

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?

Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Will the media formulations containing GlutaMAX supplement change with respect to L-glutamine content?

In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.