Acetonitrile, 99.9%, for HPLC gradient grade, ACS reagent, meets the requirem. of Reag. Ph.Eur.
Acetonitrile, 99.9%, for HPLC gradient grade, ACS reagent, meets the requirem. of Reag. Ph.Eur.
Acetonitrile, 99.9%, for HPLC gradient grade, ACS reagent, meets the requirem. of Reag. Ph.Eur.
Thermo Scientific Chemicals

Acetonitrile, 99.9%, for HPLC gradient grade, ACS reagent, meets the requirem. of Reag. Ph.Eur.

Green features
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 325730025
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Filtered to 0.2 micrometerFiltered
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only