Ammonium cerium(IV) sulfate dihydrate, 95%, ACS reagent
Ammonium cerium(IV) sulfate dihydrate, 95%, ACS reagent
Ammonium cerium(IV) sulfate dihydrate, 95%, ACS reagent
Acros Organics

Ammonium cerium(IV) sulfate dihydrate, 95%, ACS reagent

CAS: 10378-47-9 | H16CeN4O16S4·2H2O | 632.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Catalog number 325791000
HSN 28461090
Price (INR)
15,495.00
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
15,495.00
Each
Chemical Identifiers
CAS10378-47-9
IUPAC Nameλ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
Molecular FormulaCeH20N4O18S4
InChI KeyVCNAMBGKEDPVGQ-UHFFFAOYSA-J
SMILESN.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Phosphate (PO4)=<0.03 %
Iron (Fe)=<0.01 %
Appearance (Color)Yellow to orange
Appearance (Form)Crystalline powder
Titration>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only