3,4-Diaminothiophene dihydrochloride, 96%
3,4-Diaminothiophene dihydrochloride, 96%
3,4-Diaminothiophene dihydrochloride, 96%
Thermo Scientific Chemicals

3,4-Diaminothiophene dihydrochloride, 96%

CAS: 90069-81-1 | C4H6N22HCl | 187.09 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 325890050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS90069-81-1
IUPAC Namethiophene-3,4-diamine dihydrochloride
Molecular FormulaC4H8Cl2N2S
InChI KeyRAMOMCXNLLLICQ-UHFFFAOYSA-N
SMILESCl.Cl.NC1=CSC=C1N
さらに表示
Appearance (Color)Grey to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only