3,4-Dichlorobenzyl bromide, 97%
3,4-Dichlorobenzyl bromide, 97%
3,4-Dichlorobenzyl bromide, 97%
Thermo Scientific Chemicals

3,4-Dichlorobenzyl bromide, 97%

CAS: 18880-04-1 | C7H5BrCl2 | 239.93 g/mol
Have Questions?
Catalog NumberQuantityPackaging
32616025025 gGlass bottle
Catalog number 326160250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS18880-04-1
IUPAC Name4-(bromomethyl)-1,2-dichlorobenzene
Molecular FormulaC7H5BrCl2
InChI KeyXLWSBDFQAJXCQX-UHFFFAOYSA-N
SMILESClC1=C(Cl)C=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to brown
Appearance (Form)Liquid
Refractive index1.6050 to 1.6080 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only