Ethyl acetate, for pesticide residue analysis
Ethyl acetate, for pesticide residue analysis
Ethyl acetate, for pesticide residue analysis
Thermo Scientific Chemicals

Ethyl acetate, for pesticide residue analysis

CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 326610010
Price (EUR)
59,30
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
59,30
Each
Chemical Identifiers
CAS141-78-6
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Residue after evaporation=<2 ppm
Appearance (Color)Clear colorless
Water=<0.05 % (Coulometric)
GC(FID) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
RUO – Research Use Only