Methanol, 99.8%, for residue analysis, ECD tested for pesticide analysis
Methanol, 99.8%, for residue analysis, ECD tested for pesticide analysis
Methanol, 99.8%, for residue analysis, ECD tested for pesticide analysis
Thermo Scientific Chemicals

Methanol, 99.8%, for residue analysis, ECD tested for pesticide analysis

CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 326630010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.0002 %
Color scale=<10 APHA
Water=<0.1 % (Coulometric)
Appearance (Form)Clear liquid
GC>=99.8 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only