Cyclohexane, 99.5%, Extra Dry, AcroSeal™
Cyclohexane, 99.5%, Extra Dry, AcroSeal™
Cyclohexane, 99.5%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

Cyclohexane, 99.5%, Extra Dry, AcroSeal™

CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 326830010
Price (USD)
91.65
Online exclusive
102.00
Save 10.35
Each
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
91.65
Online exclusive
102.00
Save 10.35
Each
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.4 %
Water=<0.005 % (Coulometric)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only