Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™
Thermo Scientific Chemicals

Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™

Dichloromethane, 99.9%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, methylene chloride, freon 30, methylene bichloride, methane, dichloro, methane dichloride, narkotil, methylene dichloride, solmethine, solaesthin, 2.5L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless to Yellow | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number 326851000
Price (USD)
56.65
Online Exclusive
62.40
Save 5.75 (9%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
62.40
Save 5.75 (9%)
Each
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.85 %
Water=<0.003 % (Coulometric)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only