1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™
1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™
1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™
1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™
Thermo Scientific Chemicals

1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number 326891000
Price (USD)
74.65
Online Exclusive
82.30
Save 7.65 (9%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
74.65
Online Exclusive
82.30
Save 7.65 (9%)
Each
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Peroxides=<0.005 % (as H2O2)
Stabilizer2 to 5 ppm BHT
Appearance (Color)Clear colorless
GC>=99.75 %
Acidity (CH3COOH)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only