Ethyl acetate, 99.9%, Extra Dry, AcroSeal™
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

Ethyl acetate, 99.9%, Extra Dry, AcroSeal™

CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number 326900025
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS141-78-6
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Residue after evaporation=<0.0005 %
Water=<0.005 % (Coulometric)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only