Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™
Thermo Scientific Chemicals

Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™

CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
View all
Glass Bottle
Catalog number 326970025
Price (EUR)
268,65
Online Exclusive
298,00
Save 29,35 (10%)
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
268,65
Online Exclusive
298,00
Save 29,35 (10%)
Each
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=99.80 %
Water=<0.005 % (Coulometric)
Acidity (CH3COOH)=<0.0005 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only