Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and Protease free
Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and Protease free
Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals

Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and Protease free

CAS: 114456-61-0 | C6H5Na3O7 | 258.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 327162500
Price (USD)
54.60
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
54.60
Each
Chemical Identifiers
CAS114456-61-0
IUPAC Nametrisodium 2-hydroxypropane-1,2,3-tricarboxylate
Molecular FormulaC6H5Na3O7
InChI KeyHRXKRNGNAMMEHJ-UHFFFAOYSA-K
SMILES[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Assay>=99.75 % (on dry substance)
Loss on drying11 % to 13 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only