Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantityPackaging
327182500250mLGlass bottle
Catalog number 327182500
Price (EUR)
81,00
Each
Quantity:
250mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
81,00
Each
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
GC>=99.80 %
Water=<0.1 % (Coulometric)
Residue after evaporation=<0.001 %
Dnase,Rnase,Protease activitynone detected
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only