Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, 99.5%, for molecular biology, DNAse, RNAse and Protease free

CAS: 13235-36-4 | C10H20N2Na4O12 | 452.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
327211000100 gGlass bottle
Catalog number 327211000
Price (USD)
68.50
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
68.50
Each
Chemical Identifiers
CAS13235-36-4
SpecificationsSpecification SheetSpecification Sheet
Dnase,Rnase,Protease activityNot detected
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Complexometric>=99.4 %
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only