Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99+%, for molecular biology, DNAse, R

Catalog number: 327245000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99+%, for molecular biology, DNAse, R

Catalog number: 327245000
500 mg, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 327245000
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7240-90-6
IUPAC Name
X-gal
Molecular Formula
C23H41Cu2N5O5
InChI Key
GGILAJWFDJWIQO-UHFFFAOYSA-N
SMILES
[Cu+].[Cu++].CC(O)O.CC(O)O.CN1CCN(CC2=CC(=CC(CN3CCN(C)CC3)=C2O)C#N)CC1
Appearance (Color)
White
Infrared spectrum
Conforms
TLC
>=99.0 %
Specific optical rotation
-61° to -63° (20°C, 589 nm) (c=1, H2O/DMF 1/1)
Dnase,Rnase,Protease activity
none detected

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell