Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals

Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free

CAS: 7647-14-5 | ClNa | 58.44 g/mol
製品番号(カタログ番号) 327300025
価格(JPY)
-
数量:
2.5 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
さらに表示
UVat 280 nm A: =<0.01
Sulfate (SO4)=<4 ppm
Iron (Fe)=<2 ppm
Appearance (Color)White
Appearance (Form)Crystalline solid
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only