Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0
Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0
Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0
Thermo Scientific Chemicals

Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0

CAS: 38641-82-6 | C14H27N3O11 | 413.38 g/mol
Have Questions?
Catalog NumberQuantity
327345000500 mL
Catalog number 327345000
Price (USD)
113.65
Special offer
Online exclusive
Ends: 28-Dec-2026
133.00
Save 19.35
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 28-Dec-2026
133.00
Save 19.35
Each
Chemical Identifiers
CAS38641-82-6
IUPAC Name2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid; 2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC14H27N3O11
InChI KeyVLEIUWBSEKKKFX-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Dnase,Rnase,Protease activityNone detected
Appearance (Color)Clear colorless
pH7.9 to 8.1 (at 25°C)
Composition:
EDTA1 mM
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent, pH buffer
RUO – Research Use Only