Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals

Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free

CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Glass Bottle
Catalog number 327380010
Price (USD)
118.65
Special offer
Online exclusive
Ends: 28-Dec-2026
139.00
Save 20.35
Each
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
118.65
Special offer
Online exclusive
Ends: 28-Dec-2026
139.00
Save 20.35
Each
Chemical Identifiers
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<10 ppm
UVat 280 nm A: =<0.05
Chloride (Cl)=<5 ppm
Assay>=99.5 %
UV(8 M in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only