This product ships via ground from US northeast.
Water, for molecular biology, DNAse, RNAse and Protease free
Water, for molecular biology, DNAse, RNAse and Protease free
Water, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals

Water, for molecular biology, DNAse, RNAse and Protease free

Water, H2O, CAS Number-7732-18-5, dihydrogen oxide, dihydrogen monoxide, 1L, 100.0 deg.C, CHEBI:15377, 0.1ppm max., Undesignated, 1.0000g/mL, none detected, 16,383, 18.02, Molecular Biology, XLYOFNOQVPJJNP-UHFFFAOYSA-N, oxidane, 1ppb max., 15, 10237, MFCD00011332, 18.015, DNase, RNase, Protease free | CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Packaging:
View all
Plastic Drum
Catalog number 327390050
Price (USD)
245.65
Special offer
Online exclusive
Ends: 30-Jun-2026
289.00
Save 43.35 (15%)
Each
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
245.65
Special offer
Online exclusive
Ends: 30-Jun-2026
289.00
Save 43.35 (15%)
Each
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
Resistivity>=18 MOhm.cm (at manufacturing)
TOC=<50 ppb
Residue after evaporation=<0.0001 % w/w
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only