Bicine, 99+%, for biochemistry
Bicine, 99+%, for biochemistry
Bicine, 99+%, for biochemistry
Thermo Scientific Chemicals

Bicine, 99+%, for biochemistry

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 327715000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS150-25-4
IUPAC Namesodium 2-[bis(2-hydroxyethyl)amino]acetate
Molecular FormulaC6H12NNaO4
InChI KeyMFBDBXAVPLFMNJ-UHFFFAOYSA-M
SMILES[Na+].OCCN(CCO)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Water=<1 % (K.F.)
Titration with NaOH>=99.0 %
Heavy metals (as Pb)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only