n-Hexane, 97%, for electronic use (MOS), residue free
n-Hexane, 97%, for electronic use (MOS), residue free
n-Hexane, 97%, for electronic use (MOS), residue free
Thermo Scientific Chemicals

n-Hexane, 97%, for electronic use (MOS), residue free

CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3278900101 LGlass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 327890010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Acidity=<0.002 %
Chloride (Cl)=<2 ppm
Barium (Ba)=<20 ppb
Appearance (Form)Clear liquid
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only