Daidzein, 98%
Daidzein, 98%
Daidzein, 98%
Thermo Scientific Chemicals

Daidzein, 98%

CAS: 486-66-8 | C15H10O4 | 254.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Packaging:
Glass bottle
Catalog number 328230250
Price (USD)
121.00
Each
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
121.00
Each
Chemical Identifiers
CAS486-66-8
IUPAC Name7-hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
Molecular FormulaC15H10O4
InChI KeyZQSIJRDFPHDXIC-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1=COC2=CC(O)=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only