Chemical Identifiers
IUPAC Name[amino({[(4S)-4-[(2S)-2-[(2S)-2-acetamido-4-methylpentanamido]-4-methylpentanamido]-5-oxopentyl]amino})methylidene]azanium
Molecular FormulaC20H39N6O4
InChI KeyGDBQQVLCIARPGH-ULQDDVLXSA-O
SMILESCC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=[NH2+])C=O
Molecular Weight (g/mol)427.57
View more