Thapsigargin, 97%
Thapsigargin, 97%
Thapsigargin, 97%
Thermo Scientific Chemicals

Thapsigargin, 97%

CAS: 67526-95-8 | C34H50O12 | 650.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Catalog number 328570050
Price (TWD)
-
Quantity:
5 mg
Request bulk or custom format
Chemical Identifiers
CAS67526-95-8
IUPAC Name(3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-(acetyloxy)-4-(butanoyloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-{[(2Z)-2-methylbut-2-enoyl]oxy}-2-oxo-2H,3H,3aH,4H,5H,6H,6aH,7H,8H,9bH-azuleno[4,5-b]furan-7-yl octanoate
Molecular FormulaC34H50O12
InChI KeyIXFPJGBNCFXKPI-FSIHEZPISA-N
SMILESCCCCCCCC(=O)O[C@@H]1[C@@H](OC(=O)C(\C)=C/C)C(C)=C2[C@@H]3OC(=O)[C@@](C)(O)[C@@]3(O)[C@H](C[C@](C)(OC(C)=O)[C@@H]12)OC(=O)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to light yellow
Appearance (Form)Powder or film
TLC>=96.0 %
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only