N-omega-Monomethyl-L-arginine monoacetate, 99%
N-omega-Monomethyl-L-arginine monoacetate, 99%
N-omega-Monomethyl-L-arginine monoacetate, 99%
N-omega-Monomethyl-L-arginine monoacetate, 99%
Thermo Scientific Chemicals

N-omega-Monomethyl-L-arginine monoacetate, 99%

CAS: 53308-83-1 | C9H20N4O4 | 248.283 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
100 mg
Packaging:
Glass Bottle
Catalog number 328721000
Price (EUR)
395,00
Each
Add to cart
Quantity:
100 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
395,00
Each
Add to cart
Chemical Identifiers
CAS53308-83-1
IUPAC Name(2S)-2-amino-5-(N''-methylcarbamimidamido)pentanoic acid; acetic acid
Molecular FormulaC9H20N4O4
InChI KeyIKPNWIGTWUZCKM-JEDNCBNOSA-N
SMILESCC(O)=O.CN=C(N)NCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
TLC>=98.5 %
Melting point180°C to 190°C
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only