N-omega-Nitro-L-arginine-methyl ester hydrochloride, 98%
N-omega-Nitro-L-arginine-methyl ester hydrochloride, 98%
N-omega-Nitro-L-arginine-methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

N-omega-Nitro-L-arginine-methyl ester hydrochloride, 98%

CAS: 51298-62-5 | C7H16ClN5O4 | 269.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 328750050
Price (USD)
147.65
Online exclusive
164.00
Save 16.35
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
147.65
Online exclusive
164.00
Save 16.35
Each
Chemical Identifiers
CAS51298-62-5
IUPAC Namehydrogen methyl (2S)-2-amino-5-{[amino(nitroamino)methylidene]amino}pentanoate chloride
Molecular FormulaC7H16ClN5O4
InChI KeyQBNXAGZYLSRPJK-JEDNCBNOSA-N
SMILES[H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder or crystalline solid
Infrared spectrumConforms
HPLC>=97.5 %
Specific optical rotation+14.0° to +16.0° (24°C, 589 nm) (c=3, CH3OH)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only