Nifedipine, 98+%
Nifedipine, 98+%
Nifedipine, 98+%
Thermo Scientific Chemicals

Nifedipine, 98+%

CAS: 21829-25-4 | C17H18N2O6 | 346.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 329270250
Price (EUR)
351,65
Online Exclusive
390,00
Save 38,35 (10%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
351,65
Online Exclusive
390,00
Save 38,35 (10%)
Each
Add to cart
Chemical Identifiers
CAS21829-25-4
IUPAC Name3,5-dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molecular FormulaC17H18N2O6
InChI KeyHYIMSNHJOBLJNT-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.0 %
Residue after ignition=<0.1 %
Appearance (Color)Yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only