G418 sulfate, for biochemistry
G418 sulfate, for biochemistry
G418 sulfate, for biochemistry
Thermo Scientific Chemicals

G418 sulfate, for biochemistry

G418 sulfate, geneticin, CAS # 108321-42-2, is an antibiotic closely related to Gentamicin B, the common aminoglycoside antibiotic.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
3294000101 gGlass Bottle
3294000505 gGlass Bottle
Catalog number 329400010
Price (USD)
150.00
Each
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
150.00
Each
Chemical Identifiers
CAS108321-42-2
IUPAC Name(2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
Molecular FormulaC20H44N4O18S2
InChI KeyUHEPSJJJMTWUCP-KETIWVBHNA-N
SMILESOS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Water=<10 %
Potency>=650 µg/mg
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• G418 sulfate (geneticin) is an aminoglycoside antibiotic isolated from Micromonospora rhodorangea
• G418 is a bacteriostatic that prevents polypeptide formation by inhibiting the elongation step of polypeptide synthesis


Applications

• G418 sulfate can be used for selection of genetically engineered cells in both eukaryotes and prokaryotes
• Laboratory grade for applications such as biochemistry

RUO – Research Use Only

Literature references

Arshad, H.; Patel, Z.; Mehrabian, M.; Bourkas, M.; Al-Azzawi, Z.; Schmitt-Ulms, G., & Watts, J. C. 2021. The aminoglycoside G418 hinders de novo prion infection in cultured cells. The Journal of biological chemistry, 297(3), 101073.

Ferguson, M. W.; Gerak, C.; Chow, C.; Rastelli, E. J.; Elmore, K. E.; Stahl, F.; Hosseini-Farahabadi, S.; Baradaran-Heravi, A.; Coltart, D. M., & Roberge, M. 2019. The antimalarial drug mefloquine enhances TP53 premature termination codon readthrough by aminoglycoside G418. PloS one, 14(5), e0216423.