HEPES free acid, 99+%, for molecular biology, DNase, Rnase and Protease free
HEPES free acid, 99+%, for molecular biology, DNase, Rnase and Protease free
HEPES free acid, 99+%, for molecular biology, DNase, Rnase and Protease free
Thermo Scientific Chemicals

HEPES free acid, 99+%, for molecular biology, DNase, Rnase and Protease free

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
Catalog number 329850500
Price (USD)
188.65
Online Exclusive
209.00
Save 20.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
188.65
Online Exclusive
209.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS7365-45-9
IUPAC Name2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular FormulaC8H18N2O4S
InChI KeyJKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILESOCCN1CCN(CCS(O)(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<0.5 % (105°C)
Iron (Fe)=<5 ppm
Appearance (Color)White
pH5 to 6.5 (0.5 M in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only