D(+)-Maltose monohydrate, 92+%
D(+)-Maltose monohydrate, 92+%
D(+)-Maltose monohydrate, 92+%
Thermo Scientific Chemicals

D(+)-Maltose monohydrate, 92+%

CAS: 6363-53-7 | C12H24O12 | 360.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 329915000
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS6363-53-7
IUPAC Name2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
Molecular FormulaC12H24O12
InChI KeyHBDJFVFTHLOSDW-UHFFFAOYNA-N
SMILESO.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Remaindermainly Glucose and Maltotriose
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only