4-chlorobutyronitrile, 97%
4-chlorobutyronitrile, 97%
4-chlorobutyronitrile, 97%
Acros Organics

4-chlorobutyronitrile, 97%

CAS: 628-20-6 | C4H6ClN | 103.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 329940050
also known as 32994
HSN 29269000
Price (INR)
3,768.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
3,768.00
Each
Chemical Identifiers
CAS628-20-6
IUPAC Name4-chlorobutanenitrile
Molecular FormulaC4H6ClN
InChI KeyZFCFBWSVQWGOJJ-UHFFFAOYSA-N
SMILESClCCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.4420 to 1.4440 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only