Acrylamide / N,N'-Methylenebisacrylamide 29:1, for biochemistry, 40% mix solution in water
Acrylamide / N,N'-Methylenebisacrylamide 29:1, for biochemistry, 40% mix solution in water
Acrylamide / N,N'-Methylenebisacrylamide 29:1, for biochemistry, 40% mix solution in water
Thermo Scientific Chemicals

Acrylamide / N,N'-Methylenebisacrylamide 29:1, for biochemistry, 40% mix solution in water

Acrylamide / N,N'-Methylenebisacrylamide, CAS # 79-06-1, 7732-18-5, 110-26-9, 79-06-1, is a water soluble compound that cross-links to produce polyacrylamide gels that are widely used in chromatography and the electrophoretic separation of nucleic acids and proteins. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 330215000
Price (USD)
215.00
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
215.00
Each
Chemical Identifiers
CAS79-06-1
IUPAC Nameprop-2-enamide
Molecular FormulaC3H5NO
InChI KeyHRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILESNC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
UVat 290 nm A: =<0.7
Appearance (Form)Solution
Conductivity=<100 µS/cm
UV(5% in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Acrylamide / N,N'-Methylenebisacrylamide is a water-soluble, odorless, white crystalline powder
  • This compound polymerizes in the presence of free radicals to generate gels whose pore-size may be regulated by manipulating the relative concentrations of this mixture of compounds

Applications

  • Polyacrylamide gels are widely used in biochemistry, forensic chemistry, genetics, molecular biology, and biotechnology to separate proteins or nucleic acids, according to their electrophoretic mobility
  • Because of their higher resolution compared to other gels, polyacrylamide gels are also used in the analysis of smaller RNA species from mixed samples
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Petrov, A.; Tsa, A.; Puglisi, J.D. Chapter Sixteen – Analysis of RNA by Analytical Polyacrylamide Gel Electrophoresis. Methods in Enzymology. 2013, 530, 301–313.
  2. Shapiro, A. L.; Viñuela, E.; Maizel, J. V. Molecular weight estimation of polypeptide chains by electrophoresis in SDS-polyacrylamide gels. Biochem. Biophys. Res. Commun. 1967, 28 (5), 815–20.
  3. Duchesne, L. G.; Lam, J. S.; MacDonald, L. A.; et al. Effect of pH and acrylamide concentration on the separation of lipopolysaccharides in polyacrylamide gels. Current Microbiology. 1988, 16 (4), 191–4.