Thermo Scientific™
Forma™ Steri-Cult™ CO2 Incubator, 232L & 323L, CTS Series
Catalog number: 3307CTS
Thermo Scientific™
Forma™ Steri-Cult™ CO2 Incubator, 232L & 323L, CTS Series
Catalog number: 3307CTS
Thermo Scientific™ Forma™ Steri-Cult™ CO2 Incubators offer the ultimate protection for high-value cultures with every element of their design, from the ISO Class 5 HEPA filtration system to their easy cleaning processes and proven automated sterilization cycles. As the largest capacity stackable CO2 incubator in the market today, the CTS Series supports cell therapy needs from research to high volume production, providing the documentation critical to validation every step of the way.
Catalog Number
3307CTS
Unit Size
Each
Description
Single 232 L incubator
Electrical Requirements
115 V, 50/60 Hz
Includes
Factory Acceptance Testing Pk., IQ/OQ Field Service
Price (USD)
Full specifications
Voltage115 V
Capacity (English)8.2 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 232 L incubator
Capacity (Metric)232.2 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 20.8 x 32.8 in. (52.3 x 52.8 x 83.8 cm)
Dimensions (L x W x H)27 x 35 x 39.4 in. (68.6 x 88.9 x 100.1 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements115 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk., IQ/OQ Field Service
Weight (English)330 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single 232 L incubator
Electrical Requirements
230 V, 50/60 Hz
Includes
Factory Acceptance Testing Pk., IQ/OQ Field Service
Price (USD)
Full specifications
Voltage230 V
Capacity (English)8.2 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 232 L incubator
Capacity (Metric)232.2 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 20.8 x 32.8 in. (52.3 x 52.8 x 83.8 cm)
Dimensions (L x W x H)27 x 35 x 39.4 in. (68.6 x 88.9 x 100.1 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements230 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk., IQ/OQ Field Service
Weight (English)330 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single 323 L incubator
Electrical Requirements
115 V, 50/60 Hz
Includes
Factory Acceptance Testing Pk., IQ/OQ Field Service
Price (USD)
Full specifications
Voltage115 V
Capacity (English)11.4 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 323 L incubator
Capacity (Metric)322.8 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 28.8 x 32.8 in. (52.3 x 73.2 x 83.8 cm)
Dimensions (L x W x H)27 x 43 x 39.4 in. (68.6 x 109.2 x 100.2 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements115 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk., IQ/OQ Field Service
Weight (English)410 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single 323 L incubator
Electrical Requirements
230 V, 50/60 Hz
Includes
Factory Acceptance Testing Pk., IQ/OQ Field Service
Price (USD)
Full specifications
Voltage230 V
Capacity (English)11.4 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 323 L incubator
Capacity (Metric)322.8 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 28.8 x 32.8 in. (52.3 x 73.2 x 83.8 cm)
Dimensions (L x W x H)27 x 43 x 39.4 in. (68.6 x 109.2 x 100.2 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements230 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk., IQ/OQ Field Service
Weight (English)410 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single 232 L incubator
Electrical Requirements
115 V, 50/60 Hz
Includes
Factory Acceptance Testing Pk.
Price (USD)
Full specifications
Voltage115 V
Capacity (English)8.2 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 232 L incubator
Capacity (Metric)232.2 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 20.8 x 32.8 in. (52.3 x 52.8 x 83.8 cm)
Dimensions (L x W x H)27 x 35 x 39.4 in. (68.6 x 88.9 x 100.1 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements115 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk.
Weight (English)330 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single 232 L incubator
Electrical Requirements
230 V, 50/60 Hz
Includes
Factory Acceptance Testing Pk.
Price (USD)
Full specifications
Voltage230 V
Capacity (English)8.2 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 232 L incubator
Capacity (Metric)232.2 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 20.8 x 32.8 in. (52.3 x 52.8 x 83.8 cm)
Dimensions (L x W x H)27 x 35 x 39.4 in. (68.6 x 88.9 x 100.1 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements230 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk.
Weight (English)330 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single 323 L incubator
Electrical Requirements
115 V, 50/60 Hz
Includes
Factory Acceptance Testing Pk.
Price (USD)
Full specifications
Voltage115 V
Capacity (English)11.4 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 323 L incubator
Capacity (Metric)322.8 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 28.8 x 32.8 in. (52.3 x 73.2 x 83.8 cm)
Dimensions (L x W x H)27 x 43 x 39.4 in. (68.6 x 109.2 x 100.2 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements115 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk.
Weight (English)410 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single 323 L incubator
Electrical Requirements
230 V, 50/60 Hz
Includes
Factory Acceptance Testing Pk.
Price (USD)
Full specifications
Voltage230 V
Capacity (English)11.4 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 323 L incubator
Capacity (Metric)322.8 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 28.8 x 32.8 in. (52.3 x 73.2 x 83.8 cm)
Dimensions (L x W x H)27 x 43 x 39.4 in. (68.6 x 109.2 x 100.2 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements230 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk.
Weight (English)410 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
Showing 8 of 8
Catalog NumberSpecificationsUnit SizeDescriptionElectrical RequirementsIncludesPrice (USD)
3307CTSFull specifications
EachSingle 232 L incubator115 V, 50/60 HzFactory Acceptance Testing Pk., IQ/OQ Field ServiceRequest A Quote
Voltage115 V
Capacity (English)8.2 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 232 L incubator
Capacity (Metric)232.2 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 20.8 x 32.8 in. (52.3 x 52.8 x 83.8 cm)
Dimensions (L x W x H)27 x 35 x 39.4 in. (68.6 x 88.9 x 100.1 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements115 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk., IQ/OQ Field Service
Weight (English)330 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
3308CTSFull specifications
EachSingle 232 L incubator230 V, 50/60 HzFactory Acceptance Testing Pk., IQ/OQ Field ServiceRequest A Quote
Voltage230 V
Capacity (English)8.2 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 232 L incubator
Capacity (Metric)232.2 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 20.8 x 32.8 in. (52.3 x 52.8 x 83.8 cm)
Dimensions (L x W x H)27 x 35 x 39.4 in. (68.6 x 88.9 x 100.1 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements230 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk., IQ/OQ Field Service
Weight (English)330 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
3310CTSFull specifications
EachSingle 323 L incubator115 V, 50/60 HzFactory Acceptance Testing Pk., IQ/OQ Field ServiceRequest A Quote
Voltage115 V
Capacity (English)11.4 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 323 L incubator
Capacity (Metric)322.8 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 28.8 x 32.8 in. (52.3 x 73.2 x 83.8 cm)
Dimensions (L x W x H)27 x 43 x 39.4 in. (68.6 x 109.2 x 100.2 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements115 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk., IQ/OQ Field Service
Weight (English)410 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
3311CTSFull specifications
EachSingle 323 L incubator230 V, 50/60 HzFactory Acceptance Testing Pk., IQ/OQ Field ServiceRequest A Quote
Voltage230 V
Capacity (English)11.4 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 323 L incubator
Capacity (Metric)322.8 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 28.8 x 32.8 in. (52.3 x 73.2 x 83.8 cm)
Dimensions (L x W x H)27 x 43 x 39.4 in. (68.6 x 109.2 x 100.2 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements230 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk., IQ/OQ Field Service
Weight (English)410 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
3307EFull specifications
EachSingle 232 L incubator115 V, 50/60 HzFactory Acceptance Testing Pk.Request A Quote
Voltage115 V
Capacity (English)8.2 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 232 L incubator
Capacity (Metric)232.2 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 20.8 x 32.8 in. (52.3 x 52.8 x 83.8 cm)
Dimensions (L x W x H)27 x 35 x 39.4 in. (68.6 x 88.9 x 100.1 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements115 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk.
Weight (English)330 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
3308EFull specifications
EachSingle 232 L incubator230 V, 50/60 HzFactory Acceptance Testing Pk.Request A Quote
Voltage230 V
Capacity (English)8.2 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 232 L incubator
Capacity (Metric)232.2 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 20.8 x 32.8 in. (52.3 x 52.8 x 83.8 cm)
Dimensions (L x W x H)27 x 35 x 39.4 in. (68.6 x 88.9 x 100.1 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements230 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk.
Weight (English)330 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
3310EFull specifications
EachSingle 323 L incubator115 V, 50/60 HzFactory Acceptance Testing Pk.Request A Quote
Voltage115 V
Capacity (English)11.4 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 323 L incubator
Capacity (Metric)322.8 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 28.8 x 32.8 in. (52.3 x 73.2 x 83.8 cm)
Dimensions (L x W x H)27 x 43 x 39.4 in. (68.6 x 109.2 x 100.2 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements115 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk.
Weight (English)410 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
3311EFull specifications
EachSingle 323 L incubator230 V, 50/60 HzFactory Acceptance Testing Pk.Request A Quote
Voltage230 V
Capacity (English)11.4 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data Outputs4-20 mA
DescriptionSingle 323 L incubator
Capacity (Metric)322.8 L
Chamber MaterialElectropolished Stainless Steel
Dimensions (D x W x H) Interior20.6 x 28.8 x 32.8 in. (52.3 x 73.2 x 83.8 cm)
Dimensions (L x W x H)27 x 43 x 39.4 in. (68.6 x 109.2 x 100.2 cm)
Temperature Range (Metric)Ambient +5°C to 50°C
Relative Humidity95%, controllable
MaterialStainless Steel
Electrical Requirements230 V, 50/60 Hz
Humidity SourceExternal humidity reservoir
IncludesFactory Acceptance Testing Pk.
Weight (English)410 lb.
CO2 Sensor TechnologyIR Sensor
LightingLED
TypeCO2 Incubator
CO2 Concentration Range0 to 20%
Unit SizeEach
Showing 8 of 8
Thermo Scientific™ Forma™ Steri-Cult™ CO2 incubators are designed to meet the needs of today’s rapid-fire discoveries with updates that perform reliably and with minimal contamination risk to help speed up your work. Starting with the quality you’ve come to depend on from Thermo Fisher, Steri-Cult™ incubators have added features that will take your work to the next level.
  • Ultimate contamination control: proven 12-log sterilization for high-value cultures, in-chamber HEPA filtration enabling ISO Class 5 chamber conditions, and an external humidification system
  • Largest stackable culturing capacity available ideal for high-volume culturing
  • Active humidification system and IR CO2 sensor with in-chamber fan for optimum cell viability
  • Designed for easy cleaning with electropolished 100% stainless steel inner chamber
  • Compatible with common cGMP facility protocols and clean room procedures such as STERIS vaporized hydrogen peroxide (VHP) biodecontamination technology