2,3-Dichloroquinoxaline, 97%
2,3-Dichloroquinoxaline, 97%
2,3-Dichloroquinoxaline, 97%
Thermo Scientific Chemicals

2,3-Dichloroquinoxaline, 97%

CAS: 2213-63-0 | C8H4Cl2N2 | 199.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 330830100
Price (USD)
71.65
Special offer
Online exclusive
Ends: 28-Dec-2026
84.40
Save 12.75
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 28-Dec-2026
84.40
Save 12.75
Each
Chemical Identifiers
CAS2213-63-0
IUPAC Name2,3-dichloroquinoxaline
Molecular FormulaC8H4Cl2N2
InChI KeySPSSDDOTEZKOOV-UHFFFAOYSA-N
SMILESClC1=C(Cl)N=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light brown
Appearance (Form)Needles
Infrared spectrumConforms
Melting point150°C to 155°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only