Methyl 4-(hydroxymethyl)benzoate, 98%
Methyl 4-(hydroxymethyl)benzoate, 98%
Methyl 4-(hydroxymethyl)benzoate, 98%
Thermo Scientific Chemicals

Methyl 4-(hydroxymethyl)benzoate, 98%

CAS: 6908-41-4 | C9H10O3 | 166.18 g/mol
製品番号(カタログ番号)数量
33084010010 g
製品番号(カタログ番号) 330840100
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6908-41-4
IUPAC Namemethyl 4-(hydroxymethyl)benzoate
Molecular FormulaC9H10O3
InChI KeyVBWFYEFYHJRJER-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(CO)C=C1
さらに表示
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point45°C to 50°C
GC>=97.5 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only