3-Nitro-o-phenylenediamine, 98%
3-Nitro-o-phenylenediamine, 98%
3-Nitro-o-phenylenediamine, 98%
Thermo Scientific Chemicals

3-Nitro-o-phenylenediamine, 98%

CAS: 3694-52-8 | C6H7N3O2 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 330850010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3694-52-8
IUPAC Name3-nitrobenzene-1,2-diamine
Molecular FormulaC6H7N3O2
InChI KeyIOCXBXZBNOYTLQ-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1N)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Dark red
Appearance (Form)Crystals or needles
Melting point157°C to 161°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only